missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RIN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30812-100ug
This item is not returnable.
View return policy
Description
RIN1 Polyclonal antibody specifically detects RIN1 in Human samples. It is validated for Western Blot
Specifications
| RIN1 | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Ras and Rab interactor 1, ras inhibitor 1, Ras inhibitor JC99, ras inhibitor RIN1, Ras interaction/interference protein 1 | |
| His-tagged recombinant human RIN1 protein | |
| 100 μg | |
| Signal Transduction | |
| 9610 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction