missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoGDI Antibody (S07-1D8), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00€ - 645.00€
Specifications
| Antigen | RhoGDI |
|---|---|
| Clone | S07-1D8 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677774
|
Novus Biologicals
NBP3-19914-50ul |
50 μg |
421.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673735
|
Novus Biologicals
NBP3-19914-100ul |
100 μg |
645.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RhoGDI Monoclonal antibody specifically detects RhoGDI in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RhoGDI | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| GDIA1, MGC117248, Rho GDI 1, Rho GDP dissociation inhibitor (GDI) alpha, rho GDP-dissociation inhibitor 1, RHOGDI, Rho-GDI alpha, RHOGDI-1 | |
| A synthetic peptide of human RhoGDI (Uniprot # P52565) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S07-1D8 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 396 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title