missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RhoGDI Antibody (S07-1D8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19914-100ul
This item is not returnable.
View return policy
Beschreibung
RhoGDI Monoclonal antibody specifically detects RhoGDI in Human, Mouse, Rat samples. It is validated for Western Blot
Spezifikation
| RhoGDI | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S07-1D8 | |
| Unconjugated | |
| GDIA1, MGC117248, Rho GDI 1, Rho GDP dissociation inhibitor (GDI) alpha, rho GDP-dissociation inhibitor 1, RHOGDI, Rho-GDI alpha, RHOGDI-1 | |
| A synthetic peptide of human RhoGDI (Uniprot # P52565) | |
| 100 μg | |
| Signal Transduction | |
| 396 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur