missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ret Mouse anti-Human, Clone: RET/2599, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | Ret |
|---|---|
| Clone | RET/2599 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 2 to 4 μg/mL, Protein Array |
| Applications | ELISA, Peptide Array |
Descrizione
Ret Monoclonal specifically detects Ret in Human samples. It is validated for ELISA, Protein Array.Specifica
| Ret | |
| 0.2 mg/mL | |
| ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
| 10 mM PBS with 0.05% BSA | |
| 5979 | |
| IgG2b κ | |
| Protein A or G purified |
| RET/2599 | |
| ELISA 2 to 4 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto