missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ret Mouse anti-Human, Clone: RET/2599, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07414-100ug
This item is not returnable.
View return policy
Description
Ret Monoclonal specifically detects Ret in Human samples. It is validated for ELISA, Protein Array.
Specifications
| Ret | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA 2 to 4 μg/mL, Protein Array | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
| 5979 | |
| Store at 4°C. | |
| IgG2b κ |
| ELISA, Peptide Array | |
| RET/2599 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction