missing translation for 'onlineSavingsMsg'
Learn More

Renalase Antibody (1C11E8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

203.00€ - 552.00€

Specifications

Antigen Renalase
Clone 1C11E8
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready
Applications Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18237750
View Documents
Novus Biologicals
NBP2-52554
0.1 mg
552.00€
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18666816
View Documents
Novus Biologicals
NBP2-52554-0.025mg
0.025 mg
203.00€
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Renalase Monoclonal specifically detects Renalase in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
Specifications

Specifications

Renalase
1 mg/mL
Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry
Unconjugated
Mouse
Lipid and Metabolism
PBS with 0.05% Sodium Azide
55328
Purified recombinant fragment of human RNLS (AA: 68-242) expressed in E. Coli.
Primary
37.8 kDa
1C11E8
Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready
Monoclonal
Purified
RUO
Human
C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase
RNLS
IgG1
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.