Learn More
Renalase Antibody (1C11E8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-52554
This item is not returnable.
View return policy
Description
Renalase Monoclonal specifically detects Renalase in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
Specifications
| Renalase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
| C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase | |
| Mouse | |
| 37.8 kDa | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 55328 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| 1C11E8 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RNLS | |
| Purified recombinant fragment of human RNLS (AA: 68-242) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |