Learn More
enQuireBio™ Recombinant Xenopus TGF-beta 1 / TGFB1 Protein
A DNA sequence encoding the Xenopus laevis (African clawed frog) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.
352.00€ - 3175.00€
Specifications
Accession Number | NP_001081330.1 |
---|---|
Gene ID (Entrez) | 397778 |
Name | TGF-beta 1 / TGFB1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075800
|
enQuireBio™
QP6781-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
16015810
|
enQuireBio™
QP6781-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
16085800
|
enQuireBio™
QP6781-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16005810
|
enQuireBio™
QP6781-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16025810
|
enQuireBio™
QP6781-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16095800
|
enQuireBio™
QP6781-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001081330.1 | |
TGF-beta 1 / TGFB1 Protein | |
Not determined. | |
Full Length | |
Xenopus | |
His | |
Tris-based buffer, 50% glycerol |
397778 | |
Research Use Only | |
TGFB1 | |
Recombinant Protein | |
Yeast | |
GVGQEYCFGN NGPNCCVKPL YINFRKDLGW KWIHEPKGYE ANYCLGNCPY IWSMDTQYSK VLSLYNQNNP GASISPCCVP DVLEPLPIIY YVGRTAKVEQ LSNMVVRSCN CS | |
Greater than 90% as determined by SDS-PAGE. |