missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Xenopus TGF-beta 1 / TGFB1 Protein

A DNA sequence encoding the Xenopus laevis (African clawed frog) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6781-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095800

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001081330.1
TGF-beta 1 / TGFB1 Protein
Research Use Only
TGFB1
Recombinant Protein
Yeast
GVGQEYCFGN NGPNCCVKPL YINFRKDLGW KWIHEPKGYE ANYCLGNCPY IWSMDTQYSK VLSLYNQNNP GASISPCCVP DVLEPLPIIY YVGRTAKVEQ LSNMVVRSCN CS
Greater than 90% as determined by SDS-PAGE.
397778
1 mg
Not determined.
Full Length
Xenopus
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers