missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae Alpha-galactosidase 1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Alpha-galactosidase 1, was expressed in the hosts and tags indicated.
352.00€ - 4075.00€
Specifications
Name | S. cerevisiae Alpha-galactosidase 1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | MEL1 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16098020
|
enQuireBio™
QP7286-YE-100UG |
100 μg |
1100.00€
100µg |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
16018030
|
enQuireBio™
QP7286-YE-1MG |
1 mg |
4075.00€
1mg |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
16008030
|
enQuireBio™
QP7286-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16028030
|
enQuireBio™
QP7286-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16038030
|
enQuireBio™
QP7286-YE-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048030
|
enQuireBio™
QP7286-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
S. cerevisiae Alpha-galactosidase 1 Protein | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
MEL1 | |
Recombinant Protein | |
Yeast | |
VSPSYNGLGL TPQMGWDNWN TFACDVSEQL LLDTADRISD LGLKDMGYKY IILDDCWSSG RDSDGFLVAD EQKFPNGMGH VADHLHNNSF LFGMYSSAGE YTCAGYPGSL GREEEDAQFF ANNRVDYLKY DNCYNKGQFG TPEISYHRYK AMSDALNKTG RPIFYSLCNW GQDLTFYWGS GIANSWRMSG DVTAEFTRPD SRCPCDGDEY DCKYAGFHCS IMNILNKAAP MGQNAGVGGW NDLDNLEVGV GNLTDDEEKA HFSMWAMVKS PLIIGANVNN LKASSYSIYS QASVIAINQD SNGIPATRVW RYYVSDTDEY GQGEIQMWSG PLDNGDQVVA LLNGGSVSRP MNTTLEEIFF DSNLGSKKLT STWDIYDLWA NRVDNSTASA ILGRNKTATG ILYNATEQSY KDGLSKNDTR LFGQKIGSLS PNAILNTTVP AHGIAFYRLR PSS | |
Greater than 90% as determined by SDS-PAGE. |