missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LOXL2 / Lysyl oxidase homolog 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) LOXL2 / Lysyl oxidase homolog 2, was expressed in the hosts and tags indicated.
245.00€ - 2645.00€
Specifications
Accession Number | NP_201582.2 |
---|---|
Gene ID (Entrez) | 94352 |
Name | LOXL2 / Lysyl oxidase homolog 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16092430
|
enQuireBio™
QP6317-YE-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
16032440
|
enQuireBio™
QP6317-YE-500UG |
500 μg |
2195.00€
500µg |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
16002440
|
enQuireBio™
QP6317-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16022440
|
enQuireBio™
QP6317-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16042440
|
enQuireBio™
QP6317-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16012440
|
enQuireBio™
QP6317-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_201582.2 | |
LOXL2 / Lysyl oxidase homolog 2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
94352 | |
Research Use Only | |
Loxl2 | |
Recombinant Protein | |
Yeast | |
QYEGWPYQLQ YPEYFQQPAP EHHQRQVPSD VVKIQVRLAG QKRKHNEGRV EVYYEGQWGT VCDDDFSIHA AHVVCRQVGY VEAKSWAASS SYGPGEGPIW LDNIYCTGKE STLASCSSNG WGVTDCKHTE DVGVVCSEKR IPGFKFDNSL INQIESLNIQ VEDIRIRPIL SAFRHRKPVT EGYVEVKEGK AWKQICNKHW TAKNSHVVCG MFGFPAEKTY NPKAYKTFAS RRKLRYWKFS MNCTGTEAHI SSCKLGPSVT RDPVKNATCE NGQPAVVSCV PSQIFSPDGP SRFRKAYKPE QPLVRLRGGA QVGEGRVEVL KNGEWGTICD DKWDLVSASV VCRELGFGTA KEAITGSRLG QGIGPIHLNE VQCTGTEKSI IDCKFNTESQ GCNHEEDAGV RCNIPIMGFQ KKVRLNGGRN PYEGRVEVLT ERNGSLVWGT VCGQNWGIVE AMVVCRQLGL GFASNAFQET WYWHGNIFAN NVVMSGVKCS GTELSLAHCR HDEEVACPEG GVRFGAGVAC SETAPDLVLN AEIVQQTAYL EDRPMSLLQC AMEENCLSAS AVHTDPTRGH RRLLRFSSQI HNNGQSDFRP KNGRHAWIWH DCHRHYHSME VFTYYDLLSL NGTKVAEGHK ASFCLEDTEC EGDIQKSYEC ANFGEQGITM GCWDMYRHDI DCQWIDITDV PPGDYLFQVV INPNYEVPES DFSNNIMKCR SRYDGYRIWM YNCHVGGAFS EETEQKFEHF SGLLNNQLSV Q | |
Greater than 90% as determined by SDS-PAGE. |