missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LOXL2 / Lysyl oxidase homolog 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) LOXL2 / Lysyl oxidase homolog 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6317-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_201582.2 | |
LOXL2 / Lysyl oxidase homolog 2 Protein | |
Research Use Only | |
Loxl2 | |
Recombinant Protein | |
Yeast | |
QYEGWPYQLQ YPEYFQQPAP EHHQRQVPSD VVKIQVRLAG QKRKHNEGRV EVYYEGQWGT VCDDDFSIHA AHVVCRQVGY VEAKSWAASS SYGPGEGPIW LDNIYCTGKE STLASCSSNG WGVTDCKHTE DVGVVCSEKR IPGFKFDNSL INQIESLNIQ VEDIRIRPIL SAFRHRKPVT EGYVEVKEGK AWKQICNKHW TAKNSHVVCG MFGFPAEKTY NPKAYKTFAS RRKLRYWKFS MNCTGTEAHI SSCKLGPSVT RDPVKNATCE NGQPAVVSCV PSQIFSPDGP SRFRKAYKPE QPLVRLRGGA QVGEGRVEVL KNGEWGTICD DKWDLVSASV VCRELGFGTA KEAITGSRLG QGIGPIHLNE VQCTGTEKSI IDCKFNTESQ GCNHEEDAGV RCNIPIMGFQ KKVRLNGGRN PYEGRVEVLT ERNGSLVWGT VCGQNWGIVE AMVVCRQLGL GFASNAFQET WYWHGNIFAN NVVMSGVKCS GTELSLAHCR HDEEVACPEG GVRFGAGVAC SETAPDLVLN AEIVQQTAYL EDRPMSLLQC AMEENCLSAS AVHTDPTRGH RRLLRFSSQI HNNGQSDFRP KNGRHAWIWH DCHRHYHSME VFTYYDLLSL NGTKVAEGHK ASFCLEDTEC EGDIQKSYEC ANFGEQGITM GCWDMYRHDI DCQWIDITDV PPGDYLFQVV INPNYEVPES DFSNNIMKCR SRYDGYRIWM YNCHVGGAFS EETEQKFEHF SGLLNNQLSV Q | |
Greater than 90% as determined by SDS-PAGE. |
94352 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |