Learn More
enQuireBio™ Recombinant Macaque IL2 / Interleukin-2 Protein
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL2 / Interleukin-2, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_001040595.1 |
---|---|
Gene ID (Entrez) | 708017 |
Name | IL2 / Interleukin-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051660
|
enQuireBio™
QP6217-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
16071660
|
enQuireBio™
QP6217-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
16061660
|
enQuireBio™
QP6217-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16081660
|
enQuireBio™
QP6217-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16091660
|
enQuireBio™
QP6217-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16001670
|
enQuireBio™
QP6217-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001040595.1 | |
IL2 / Interleukin-2 Protein | |
Not determined. | |
Full Length of Mature Protein | |
Macaque | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
708017 | |
Research Use Only | |
IL2 | |
Recombinant Protein | |
E. coli | |
APTSSSTKKT QLQLEHLLLD LQMILNGINN YKNPKLTRML TFKFYMPKKA TELKHLQCLE EELKPLEEVL NLAQSKNFHL RDTKDLISNI NVIVLELKGS ETTLMCEYAD ETATIVEFLN RWITFCQSII STLT | |
Greater than 82.1% as determined by SDS-PAGE. |