Learn More
enQuireBio™ Recombinant Macaque IL2 / Interleukin-2 Protein
Product Code. 16071660
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16071660 1 mg 1mg
16061660 10 μg 10µg
16001670 50 μg 50µg
16051660 100 μg 100µg
16081660 200 μg 200µg
16091660 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16071660 Supplier enQuireBio™ Supplier No. QP6217ec1mg
A DNA sequence encoding the Macaca mulatta (Rhesus macaque) IL2 / Interleukin-2, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_001040595.1
Gene ID (Entrez) 708017
Name IL2 / Interleukin-2 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol IL2
Protein Length Full Length of Mature Protein
Product Type Recombinant Protein
Cross Reactivity Macaque
Species E. coli
Protein Tag His-SUMO
Sequence APTSSSTKKT QLQLEHLLLD LQMILNGINN YKNPKLTRML TFKFYMPKKA TELKHLQCLE EELKPLEEVL NLAQSKNFHL RDTKDLISNI NVIVLELKGS ETTLMCEYAD ETATIVEFLN RWITFCQSII STLT
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 82.1% as determined by SDS-PAGE.