Learn More
enQuireBio™ Recombinant Human Scrapie-responsive protein 1
A DNA sequence encoding the Homo sapiens (Human) Scrapie-responsive protein 1, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Accession Number | NP_001316526.1 |
---|---|
Gene ID (Entrez) | 11341 |
Name | Scrapie-responsive protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16089020
|
enQuireBio™
QP7501-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
16029030
|
enQuireBio™
QP7501-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
16099020
|
enQuireBio™
QP7501-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16019030
|
enQuireBio™
QP7501-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16039030
|
enQuireBio™
QP7501-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16009030
|
enQuireBio™
QP7501-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001316526.1 | |
Scrapie-responsive protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
11341 | |
Research Use Only | |
SCRG1 | |
Recombinant Protein | |
Yeast | |
MPANRLSCYR KILKDHNCHN LPEGVADLTQ IDVNVQDHFW DGKGCEMICY CNFSELLCCP KDVFFGPKIS FVIPCNNQ | |
Greater than 90% as determined by SDS-PAGE. |