missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Scrapie-responsive protein 1

A DNA sequence encoding the Homo sapiens (Human) Scrapie-responsive protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7501-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16009030

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001316526.1
Scrapie-responsive protein 1
Research Use Only
SCRG1
Recombinant Protein
Yeast
MPANRLSCYR KILKDHNCHN LPEGVADLTQ IDVNVQDHFW DGKGCEMICY CNFSELLCCP KDVFFGPKIS FVIPCNNQ
Greater than 90% as determined by SDS-PAGE.
11341
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers