Learn More
ZNF268 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94146-0.1ml
This item is not returnable.
View return policy
Description
ZNF268 Polyclonal antibody specifically detects ZNF268 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ZNF268 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| zinc finger protein 446, Zinc finger protein with KRAB and SCAN domains 20, ZKSCAN20FLJ20626, ZSCAN30 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ZNF268 (NP_001159353.1). MATRVRTASIWVPPLQERNSSWDRIRKLQGQESILGQGTPGLQPLPGTPRQKQKSRRIEKVLEWLFISQEQPKITKSWGPLSFMDVFVDFTWEEWQLLDPAQKCLYRSVMLENYSNLVSLGYQHTKPDIIFKLEQGEELCMVQAQVPNQTCPNTVWKIDDLMDWHQENKDKLGSTAKSFECTTFGKLCLLSTKYLSRQKPHKCGTHGKSLKYIDFTSDYARNNPNGFQVHGKSFFHSKHEQTVIGIKYCESIESGKTVNKKSQLMCQQMY | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10795 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |