missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF268 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00€ - 470.00€
Specifications
| Antigen | ZNF268 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680272
|
Novus Biologicals
NBP2-94146-0.02ml |
0.02 mL |
188.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18676742
|
Novus Biologicals
NBP2-94146-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF268 Polyclonal antibody specifically detects ZNF268 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ZNF268 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| zinc finger protein 446, Zinc finger protein with KRAB and SCAN domains 20, ZKSCAN20FLJ20626, ZSCAN30 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human ZNF268 (NP_001159353.1). MATRVRTASIWVPPLQERNSSWDRIRKLQGQESILGQGTPGLQPLPGTPRQKQKSRRIEKVLEWLFISQEQPKITKSWGPLSFMDVFVDFTWEEWQLLDPAQKCLYRSVMLENYSNLVSLGYQHTKPDIIFKLEQGEELCMVQAQVPNQTCPNTVWKIDDLMDWHQENKDKLGSTAKSFECTTFGKLCLLSTKYLSRQKPHKCGTHGKSLKYIDFTSDYARNNPNGFQVHGKSFFHSKHEQTVIGIKYCESIESGKTVNKKSQLMCQQMY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 10795 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title