missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHB Mouse anti-Human, Clone: SDHB/2126, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07786-100ug
This item is not returnable.
View return policy
Description
SDHB Monoclonal specifically detects SDHB in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| SDHB | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 0.5-1.0 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip) | |
| Recombinant human SDHB fragment around aa 165-273 (Exact sequence is proprietary) | |
| 100 μg | |
| Cancer | |
| 6390 | |
| Store at 4°C. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| SDHB/2126 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto