missing translation for 'onlineSavingsMsg'
Learn More

SDHB Mouse anti-Human, Clone: SDHB/2126, Novus Biologicals™

Mouse Monoclonal Antibody

484.00€

Specifications

Antigen SDHB
Clone SDHB/2126
Concentration 0.2 mg/mL
Dilution Western Blot 0.5-1.0 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18635906
View Documents
Novus Biologicals
NBP3-07786-100ug
100 μg
484.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SDHB Monoclonal specifically detects SDHB in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

SDHB
0.2 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
6390
IgG1 κ
Protein A or G purified
SDHB/2126
Western Blot 0.5-1.0 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
Human
EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip)
Recombinant human SDHB fragment around aa 165-273 (Exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.