missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coagulation Factor V (Light Chain) Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9856-SP
This item is not returnable.
View return policy
Description
Factor V Monoclonal specifically detects Factor V in Human samples. It is validated for ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| FactorV | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| Activated protein C cofactor, Cf5, Cf-5, Coagulation Factor 5, coagulation factor V, coagulation factor V (proaccelerin, labile factor), coagulation factor V jinjiang A2 domain, F5, Factor V, factor V Leiden, FVL, PCCF, Proaccelerin, labile factor, RPRGL1, THPH2 | |
| Synthetic peptide containing human Coagulation Factor V aa1650-1700, Accession # P12259 | |
| 25 μg | |
| 2153 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| ELISA, Flow Cytometry, CyTOF | |
| 2280A | |
| ELISA, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P12259 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Coagulation Factor V in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction