missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coagulation Factor V (Light Chain) Antibody, R&D Systems™
Rabbit Monoclonal Antibody
183.00€ - 389.00€
Specifications
| Antigen | FactorV |
|---|---|
| Clone | 2280A |
| Dilution | ELISA, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | ELISA, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18176141
|
R&D Systems
MAB9856-SP |
25 μg |
183.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18172931
|
R&D Systems
MAB9856-100 |
100 μg |
389.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Factor V Monoclonal specifically detects Factor V in Human samples. It is validated for ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| FactorV | |
| ELISA, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P12259 | |
| 2153 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2280A | |
| ELISA, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| Activated protein C cofactor, Cf5, Cf-5, Coagulation Factor 5, coagulation factor V, coagulation factor V (proaccelerin, labile factor), coagulation factor V jinjiang A2 domain, F5, Factor V, factor V Leiden, FVL, PCCF, Proaccelerin, labile factor, RPRGL1, THPH2 | |
| Synthetic peptide containing human Coagulation Factor V aa1650-1700, Accession # P12259 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human Coagulation Factor V in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title