Learn More
enQuireBio™ Recombinant Human Angiopoietin 1 / ANG1 / ANGPT1 Protein
A DNA sequence encoding the Homo sapiens (Human) Angiopoietin 1 / ANG1 / ANGPT1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_001137.2 |
---|---|
Gene ID (Entrez) | 284 |
Name | Angiopoietin 1 / ANG1 / ANGPT1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009670
|
enQuireBio™
QP7600-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
16029670
|
enQuireBio™
QP7600-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
16019670
|
enQuireBio™
QP7600-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16039670
|
enQuireBio™
QP7600-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16049670
|
enQuireBio™
QP7600-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16059670
|
enQuireBio™
QP7600-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001137.2 | |
Angiopoietin 1 / ANG1 / ANGPT1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
284 | |
Research Use Only | |
Angpt1 | |
Recombinant Protein | |
E. coli | |
HIGCSNQRRS PENSGRRYNR IQHGQCAYTF ILPEHDGNCR ESTTDQYNTN ALQRDAPHVE PDFSSQKLQH LEHVMENYTQ WLQKLENYIV ENMKSEMAQI QQNAVQNHTA TMLEIGTSLL SQTAEQTRKL TDVETQVLNQ TSRLEIQLLE NSLSTYKLEK QLLQQTNEIL KIHEKNSLLE HKILEMEGKH KEELDTLKEE KENLQGLVTR QTYIIQELEK QLNRATTNNS VLQKQQLELM DTVHNLVNLC TKEGVLLKGG KREEEKPFRD CADVYQAGFN KSGIYTIYIN NMPEPKKVFC NMDVNGGGWT VIQHREDGSL DFQRGWKEYK MGFGNPSGEY WLGNEFIFAI TSQRQYMLRI ELMDWEGNRA YSQYDRFHIG NEKQNYRLYL KGHTGTAGKQ SSLILHGADF STKDADNDNC MCKCALMLTG GWWFDACGPS NLNGMFYTAG QNHGKLNGIK WHYFKGPSYS LRSTTMMIRP LDF | |
Greater than 90% as determined by SDS-PAGE. |