missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Angiopoietin 1 / ANG1 / ANGPT1 Protein

A DNA sequence encoding the Homo sapiens (Human) Angiopoietin 1 / ANG1 / ANGPT1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7600-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039670

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001137.2
Angiopoietin 1 / ANG1 / ANGPT1 Protein
Research Use Only
Angpt1
Recombinant Protein
E. coli
HIGCSNQRRS PENSGRRYNR IQHGQCAYTF ILPEHDGNCR ESTTDQYNTN ALQRDAPHVE PDFSSQKLQH LEHVMENYTQ WLQKLENYIV ENMKSEMAQI QQNAVQNHTA TMLEIGTSLL SQTAEQTRKL TDVETQVLNQ TSRLEIQLLE NSLSTYKLEK QLLQQTNEIL KIHEKNSLLE HKILEMEGKH KEELDTLKEE KENLQGLVTR QTYIIQELEK QLNRATTNNS VLQKQQLELM DTVHNLVNLC TKEGVLLKGG KREEEKPFRD CADVYQAGFN KSGIYTIYIN NMPEPKKVFC NMDVNGGGWT VIQHREDGSL DFQRGWKEYK MGFGNPSGEY WLGNEFIFAI TSQRQYMLRI ELMDWEGNRA YSQYDRFHIG NEKQNYRLYL KGHTGTAGKQ SSLILHGADF STKDADNDNC MCKCALMLTG GWWFDACGPS NLNGMFYTAG QNHGKLNGIK WHYFKGPSYS LRSTTMMIRP LDF
Greater than 90% as determined by SDS-PAGE.
284
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers