missing translation for 'onlineSavingsMsg'
Learn More

RAP1A Antibody (5F8G2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

203.00€ - 552.00€

Specifications

Antigen RAP1A
Clone 5F8G2
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready
Applications Western Blot, Flow Cytometry, ELISA, CyTOF
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18673746
View Documents
Novus Biologicals
NBP2-37495-0.025ml
0.025 mL
203.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18167537
View Documents
Novus Biologicals
NBP2-37495
0.1 mL
552.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RAP1A Monoclonal specifically detects RAP1A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications

Specifications

RAP1A
1 mg/mL
Western Blot, Flow Cytometry, ELISA, CyTOF
Unconjugated
RUO
Human
C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21
RAP1A
IgG1
Affinity Purified
5F8G2
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready
Monoclonal
Mouse
Cell Cycle and Replication, Endosome Markers
PBS with 0.05% Sodium Azide
5906
Purified recombinant fragment of human RAP1A expressed in E. Coli.
Primary
43.2 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.