missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP1A Antibody (5F8G2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37495-0.025ml
This item is not returnable.
View return policy
Description
RAP1A Monoclonal specifically detects RAP1A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.
Specifications
| RAP1A | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21 | |
| Mouse | |
| 43.2 kDa | |
| 0.025 mL | |
| Cell Cycle and Replication, Endosome Markers | |
| 5906 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 5F8G2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RAP1A | |
| Purified recombinant fragment of human RAP1A expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction