missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Rat IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Rabbit Polyclonal Antibody
150.00€
Specifications
| Antigen | IgG (H+L) |
|---|---|
| Concentration | 4.5 mg/mL |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.Specifications
| IgG (H+L) | |
| Immunoassay | |
| Unconjugated | |
| RUO | |
| Rat | |
| IgG | |
| Affinity purified | |
| Based on IEP, this antibody reacts with heavy gamma chains on rat IgG and light chains on all rat immunoglobulins. This antibody has been pre-adsorbed against human serum proteins. |
| 4.5 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Immunology | |
| Purified Rat IgG (H&L) | |
| Secondary | |
| Store at 4°C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title