Learn More
Rabbit anti-Rat IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Concentration | 4.5 mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Purified Rat IgG (H&L) |
| Purification Method | Affinity purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Show More |