missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3GAP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 470.00€
Specifikationer
| Antigen | RAB3GAP2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|
18693851
|
Novus Biologicals
NBP2-93097-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615440
|
Novus Biologicals
NBP2-93097-0.1ml |
0.1 mL |
470.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
RAB3GAP2 Polyclonal antibody specifically detects RAB3GAP2 in Human, Rat samples. It is validated for Western BlotSpecifikationer
| RAB3GAP2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, GPCR, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 25782 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| DKFZP434D245, FLJ14579, KIAA0839p150, RAB3 GTPase activating protein subunit 2 (non-catalytic), Rab3 GTPase-activating protein 150 kDa subunit, rab3 GTPase-activating protein non-catalytic subunit, Rab3-GAP p150, Rab3-GAP regulatory subunit, Rab3-GAP150, RAB3GAP150, RGAP-iso | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human RAB3GAP2 (NP_036546.2). MACSIVQFCYFQDLQAARDFLFPHLREEILSGALRRDPSKSTDWEDDGWGAWEENEPQEPEEEGNTCKTQKTSWLQDCVLSLSPTNDLMVIAREQKAVFLVPKWKYSDKGKEEMQFAVGWSGSLNVEEGECVTSALCIPLASQKRSSTGRPDWTCIVVGFTSGYVRFYTENGVLLLAQLLNEDPVLQLKCRTYEIPRHPG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel