missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3GAP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93097-0.1ml
This item is not returnable.
View return policy
Description
RAB3GAP2 Polyclonal antibody specifically detects RAB3GAP2 in Human, Rat samples. It is validated for Western Blot
Specifications
| RAB3GAP2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DKFZP434D245, FLJ14579, KIAA0839p150, RAB3 GTPase activating protein subunit 2 (non-catalytic), Rab3 GTPase-activating protein 150 kDa subunit, rab3 GTPase-activating protein non-catalytic subunit, Rab3-GAP p150, Rab3-GAP regulatory subunit, Rab3-GAP150, RAB3GAP150, RGAP-iso | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human RAB3GAP2 (NP_036546.2). MACSIVQFCYFQDLQAARDFLFPHLREEILSGALRRDPSKSTDWEDDGWGAWEENEPQEPEEEGNTCKTQKTSWLQDCVLSLSPTNDLMVIAREQKAVFLVPKWKYSDKGKEEMQFAVGWSGSLNVEEGECVTSALCIPLASQKRSSTGRPDWTCIVVGFTSGYVRFYTENGVLLLAQLLNEDPVLQLKCRTYEIPRHPG | |
| 0.1 mL | |
| Cell Biology, GPCR, Neuroscience, Signal Transduction | |
| 25782 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction