Learn More
Pyridazine-3-carboxylic acid, 97%
Product Code. 15457277
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457277 5 g 5g
15447277 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15457277 Supplier Thermo Scientific Alfa Aesar Supplier No. H34341.06
Request Bulk or Custom Format
CAS: 2164-61-6 | C5H4N2O2 | 124.099 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2164-61-6
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.099
MDL Number MFCD01646333
InChI Key RUUOPSRRIKJHNH-UHFFFAOYSA-N
Synonym 3-pyridazinecarboxylic acid, 3-carboxypyridazine, 3-pyridazine carboxylic acid, pubchem22109, pyridazine carboxylic acid, ksc202q0p, pyridazine-3-carboxylic acid, 3-carboxypyridazine; 3-pyridazinecarboxylic acid;
PubChem CID 269369
IUPAC Name pyridazine-3-carboxylic acid
SMILES C1=CC(=NN=C1)C(=O)O
Specifications
Melting Point ∽195°C (decomposition)
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 124.1
Percent Purity 97%
Chemical Name or Material Pyridazine-3-carboxylic acid