missing translation for 'onlineSavingsMsg'
Learn More

PSGL-1/CD162 Mouse anti-Human, Clone: PSGL1/1567, Novus Biologicals™

Mouse Monoclonal Antibody

484.00€

Specifications

Antigen PSGL-1/CD162
Clone PSGL1/1567
Concentration 0.2 mg/mL
Dilution Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL
Applications Flow Cytometry, Immunofluorescence
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18654938
View Documents
Novus Biologicals
NBP3-07820-100ug
100 μg
484.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PSGL-1/CD162
0.2 mg/mL
Flow Cytometry, Immunofluorescence
Unconjugated
Mouse
Cholesterol Metabolism, Immunology, Lipid and Metabolism
10 mM PBS with 0.05% BSA
6404
IgM κ
Column Chromatography
PSGL1/1567
Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL
Monoclonal
Purified
RUO
Human
CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA
Recombinant human PSGL-1/CD162 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.