missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSENEN/PEN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
199.00€ - 351.00€
Specifications
| Antigen | PSENEN/PEN2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 3-7 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18100942
|
Novus Biologicals
NBP2-24611SS |
0.025 mg |
199.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18107861
|
Novus Biologicals
NBP2-24611 |
0.1 mg |
351.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSENEN/PEN2 Polyclonal specifically detects PSENEN/PEN2 in Human, Mouse, Rat, Bovine, Canine, Equine, Opossum samples. It is validated for Western Blot.Specifications
| PSENEN/PEN2 | |
| Western Blot 3-7 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Opossum | |
| Q9NZ42 | |
| 55851 | |
| A portion of amino acid 1 to 50 of human Presenilin enhancer protein 2 was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Growth and Development, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2 | |
| PSENEN | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, horse, monkey, mouse, rat and 93% homologous in cow. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title