missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSENEN/PEN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24611SS
This item is not returnable.
View return policy
Description
PSENEN/PEN2 Polyclonal specifically detects PSENEN/PEN2 in Human, Mouse, Rat, Bovine, Canine, Equine, Opossum samples. It is validated for Western Blot.
Specifications
| PSENEN/PEN2 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, horse, monkey, mouse, rat and 93% homologous in cow. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 3-7 ug/ml | |
| Q9NZ42 | |
| PSENEN | |
| A portion of amino acid 1 to 50 of human Presenilin enhancer protein 2 was used as immunogen. | |
| 0.025 mg | |
| Growth and Development, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 55851 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Opossum | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction