missing translation for 'onlineSavingsMsg'
Learn More

Proinsulin Antibody (C-PEP-01), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody

553.00€

Specifications

Antigen Proinsulin
Clone C-PEP-01
Applications ELISA
Classification Monoclonal
Conjugate mFluor Violet 450 SE
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18735022
View Documents
Novus Biologicals
NB500-413MFV450
0.1 mL
553.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Proinsulin Monoclonal antibody specifically detects Proinsulin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
Specifications

Specifications

Proinsulin
ELISA
mFluor Violet 450 SE
Mouse
Diabetes Research, Immune System Diseases, Immunology
50mM Sodium Borate
3630
IgG1
Protein A purified
C-PEP-01
Monoclonal
Purified
RUO
Human
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
C-peptide (Human) conjugated to BSA.
Primary
Store at 4°C in the dark.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.