missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proinsulin Antibody (C-PEP-01), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-413MFV450
This item is not returnable.
View return policy
Description
Proinsulin Monoclonal antibody specifically detects Proinsulin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
Specifications
| Proinsulin | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| C-peptide (Human) conjugated to BSA. | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology | |
| 3630 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| C-PEP-01 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction