missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (Alpha PR6), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
604.00€
Specifications
| Antigen | Progesterone R/NR3C3 |
|---|---|
| Clone | Alpha PR6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 450 SE |
Description
Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human, Mouse, Rat, Bovine, Chicken, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| Progesterone R/NR3C3 | |
| Western Blot | |
| mFluor Violet 450 SE | |
| Mouse | |
| Breast Cancer, Cancer, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
| 50mM Sodium Borate | |
| 5241 | |
| IgG2a | |
| Protein A purified |
| Alpha PR6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Progesterone receptor purified from chick oviduct cytosol. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title