missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Progesterone R/NR3C3 Antibody (Alpha PR6), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-2765MFV450
489.60 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Progesterone R/NR3C3 Monoclonal antibody specifically detects Progesterone R/NR3C3 in Human, Mouse, Rat, Bovine, Chicken, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Progesterone R/NR3C3 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| NR3C3PRNuclear receptor subfamily 3 group C member 3, progesterone receptor | |
| Progesterone receptor purified from chick oviduct cytosol. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Immunology, Innate Immunity, Phospho Specific, Signal Transduction | |
| 5241 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot | |
| Alpha PR6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction