missing translation for 'onlineSavingsMsg'
Learn More

PRKCSH Antibody (SR2231), Novus Biologicals™

Rabbit Monoclonal Antibody

338.00€ - 449.00€

Specifications

Antigen PRKCSH
Clone SR2231
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18694494
View Documents
Novus Biologicals
NBP3-22452-100ul
100 μg
449.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18630245
View Documents
Novus Biologicals
NBP3-22452-50ul
50 μg
338.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PRKCSH Monoclonal antibody specifically detects PRKCSH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

PRKCSH
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human
AGE-R2,80K-H protein, G19P1AGE-binding receptor 2, glucosidase 2 subunit beta, Glucosidase II subunit beta, glucosidase II, beta subunit, hepatocystin, PCLD, PKCSH, PLD1, Protein kinase C substrate 60.1 kDa protein heavy chain, protein kinase C substrate 80K-H, protein kinase C substrate, 80 Kda protein
A synthesized peptide derived from human PRKCSH (Uniprot #: P14314)
Primary
Store at -20°C. Avoid freeze/thaw cycles.
SR2231
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Signal Transduction
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
5589
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.