missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKCSH Antibody (SR2231), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22452-50ul
This item is not returnable.
View return policy
Description
PRKCSH Monoclonal antibody specifically detects PRKCSH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| PRKCSH | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR2231 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| AGE-R2,80K-H protein, G19P1AGE-binding receptor 2, glucosidase 2 subunit beta, Glucosidase II subunit beta, glucosidase II, beta subunit, hepatocystin, PCLD, PKCSH, PLD1, Protein kinase C substrate 60.1 kDa protein heavy chain, protein kinase C substrate 80K-H, protein kinase C substrate, 80 Kda protein | |
| A synthesized peptide derived from human PRKCSH (Uniprot #: P14314) | |
| 50 μg | |
| Signal Transduction | |
| 5589 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction