missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PPP2R5D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

213.00€ - 508.00€

Specifications

Antigen PPP2R5D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30067010
View Documents
Novus Biologicals™
NBP3-30776-25ug
25 μg
213.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30067127
View Documents
Novus Biologicals™
NBP3-30776-100ug
100 μg
508.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PPP2R5D Polyclonal antibody specifically detects PPP2R5D in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications

Specifications

PPP2R5D
Polyclonal
Purified
RUO
Human, Mouse, Rat
MGC2134, MGC8949, PP2A B subunit isoform B56-delta, PP2A B subunit isoform B'-delta, PP2A B subunit isoform PR61-delta, PP2A B subunit isoform R5-delta, PP2A, B subunit, B' delta isoform, PP2A, B subunit, B56 delta isoform, PP2A, B subunit, PR61 delta isoform, PP2A, B subunit, R5 delta isoform, protein phosphatase 2, regulatory subunit B', delta, regulatory subunit B (B56), delta isoform, regulatory subunit B', delta isoform, Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, deltaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform
Synthetic peptide conjugated to KLH derived from the N-terminus of PPP2R5D
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot
Unconjugated
Rabbit
Neuroscience, Wnt Signaling Pathway
Phosphate buffered saline
5528
IgG
Ammonium sulfate precipitation
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.