Learn More
Novus Biologicals™ PPP2R5D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30776-100ug
This item is not returnable.
View return policy
Description
PPP2R5D Polyclonal antibody specifically detects PPP2R5D in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| PPP2R5D | |
| Polyclonal | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| MGC2134, MGC8949, PP2A B subunit isoform B56-delta, PP2A B subunit isoform B'-delta, PP2A B subunit isoform PR61-delta, PP2A B subunit isoform R5-delta, PP2A, B subunit, B' delta isoform, PP2A, B subunit, B56 delta isoform, PP2A, B subunit, PR61 delta isoform, PP2A, B subunit, R5 delta isoform, protein phosphatase 2, regulatory subunit B', delta, regulatory subunit B (B56), delta isoform, regulatory subunit B', delta isoform, Serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, deltaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit delta isoform | |
| Synthetic peptide conjugated to KLH derived from the N-terminus of PPP2R5D | |
| 100 μg | |
| Neuroscience, Wnt Signaling Pathway | |
| 5528 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |