Learn More
Hirschmann™ Amber Glass Bottle with Screw Cap
Product Code. 10389960
Click to view available options
Capacity (Metric):
2500 mL
Unit Size:
Each
Product Code. 10389960
Brand: Hirschmann™ 9315200
Only null left
Add to basket
Add to basket
An excellent option for the storage of light-sensitive materials.
  • Screw cap closure included
  • Round shape
  • A45 thread
  • 2500mL capacity
Specifications
Capacity (Metric) 2500 mL
Color Amber
For Use With (Application) Storage of light-sensitive materials
Material Glass
Quantity 1Each
Thread Size (Metric) A45
Closure Color White
Closure Included Yes
Shape Round
Product Type Amber Bottle
Closure Material Polypropylene
Closure Type Screw Cap