missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (pmab-1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
169.00€ - 329.00€
Specifications
| Antigen | Podoplanin |
|---|---|
| Clone | pmab-1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 3 - 5 ug/mL, Flow Cytometry 0.5 ug/10^5 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18068486
|
Novus Biologicals
NBP2-03955SS |
0.025 mg |
169.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18701214
|
Novus Biologicals
NBP2-03955 |
329.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Podoplanin Monoclonal specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| Podoplanin | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rat | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| NP_034459.2 | |
| 10630 | |
| Podoplanin | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| pmab-1 | |
| Western Blot 3 - 5 ug/mL, Flow Cytometry 0.5 ug/10^5 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| PDPN | |
| IgG2a κ | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title