missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Podoplanin Antibody (pmab-1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-03955SS
This item is not returnable.
View return policy
Description
Podoplanin Monoclonal specifically detects Podoplanin in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Podoplanin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 3 - 5 ug/mL, Flow Cytometry 0.5 ug/10^5 cells, CyTOF-ready | |
| 36-KD, aggrus, Gp36, GP40, HT1A-1, hT1alpha-1, hT1alpha-2, lung type I cell membrane associated glycoprotein, lung type-I cell membrane-associated glycoprotein (T1A-2), OTS 8, OTS8, PA2.26 antigen, podoplanin, T1A, T1-alpha | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| pmab-1 | |
| Unconjugated | |
| NP_034459.2 | |
| PDPN | |
| Podoplanin | |
| 0.025 mg | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 10630 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction