missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PMS1 Antibody (CL14436), Novus Biologicals™
Mouse Monoclonal Antibody
433.00€ - 652.00€
Specifications
| Antigen | PMS1 |
|---|---|
| Clone | CL14436 |
| Concentration | .8 mg/mL |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043865
|
Novus Biologicals™
NBP3-26854-100ul |
100 μL |
652.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043484
|
Novus Biologicals™
NBP3-26854-25ul |
25 μL |
433.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PMS1 Monoclonal antibody specifically detects PMS1 in Human samples. It is validated for Western BlotSpecifications
| PMS1 | |
| .8 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| DNA Repair, Mismatch Repair, Ovarian Carcinoma Cell Markers | |
| PBS, pH 7.2, 40% glycerol | |
| 5378 | |
| IgG2b | |
| Protein A purified |
| CL14436 | |
| Western Blot 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp781M0253, FLJ98259, HNPCC3, hPMS1, human homolog of yeast mutL, mismatch repair gene PMSL1, PMS1 postmeiotic segregation increased 1 (S. cerevisiae), PMS1 protein homolog 1, PMSL1DNA mismatch repair protein PMS1, postmeiotic segregation increased (S. cerevisiae) 1, rhabdomyosarcoma antigen MU-RMS-40.10B, rhabdomyosarcoma antigen MU-RMS-40.10E | |
| This antibody was developed against a Synthetic Peptide of P54277. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title