missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PMS1 Antibody (CL14436), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26854-100ul
This item is not returnable.
View return policy
Description
PMS1 Monoclonal antibody specifically detects PMS1 in Human samples. It is validated for Western Blot
Specifications
| PMS1 | |
| Monoclonal | |
| .8 mg/mL | |
| Western Blot 1 μg/mL | |
| DKFZp781M0253, FLJ98259, HNPCC3, hPMS1, human homolog of yeast mutL, mismatch repair gene PMSL1, PMS1 postmeiotic segregation increased 1 (S. cerevisiae), PMS1 protein homolog 1, PMSL1DNA mismatch repair protein PMS1, postmeiotic segregation increased (S. cerevisiae) 1, rhabdomyosarcoma antigen MU-RMS-40.10B, rhabdomyosarcoma antigen MU-RMS-40.10E | |
| This antibody was developed against a Synthetic Peptide of P54277. Actual immunogen sequence is proprietary. | |
| 100 μL | |
| DNA Repair, Mismatch Repair, Ovarian Carcinoma Cell Markers | |
| 5378 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Western Blot | |
| CL14436 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction