missing translation for 'onlineSavingsMsg'
Learn More

PLEKHG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

483.00€

Specifications

Antigen PLEKHG2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18209452
View Documents
Novus Biologicals
NBP1-69100
100 μL
483.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PLEKHG2 Polyclonal specifically detects PLEKHG2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

PLEKHG2
Polyclonal
Rabbit
ARHGEF42, CLGFLJ38638, common-site lymphoma/leukemia guanine nucleotide exchange factor, DKFZp667J2325, FLJ00018, FLJ22458, PH domain-containing family G member 2, pleckstrin homology domain containing, family G (with RhoGef domain) member 2, pleckstrin homology domain-containing family G member 2
PLEKHG2
Primary
Western Blot
Unconjugated
RUO
64857
IgG
144 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.