missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pleiotrophin/PTN Antibody (CL15628) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
376.00€ - 561.00€
Specifications
| Antigen | Pleiotrophin/PTN |
|---|---|
| Clone | CL15628 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279555
|
Novus Biologicals
NBP3-43677-25ul |
25 μL |
376.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279293
|
Novus Biologicals
NBP3-43677-100ul |
100 μL |
561.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pleiotrophin/PTN Monoclonal antibody specifically detects Pleiotrophin/PTN in Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)Specifications
| Pleiotrophin/PTN | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| PBS, pH 7.2, 40% glycerol | |
| 5764 | |
| IgG1 | |
| Protein A purified |
| CL15628 | |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| HARP, HBBM, HB-GAM, HBGF8, HBNF, HBNF1, HBNF-1, Heparin-binding brain mitogen, NEGF1HBGF-8, neurite growth-promoting factor 1, neurite growth-promoting factor1), OSF-1, Osteoblast-specific factor 1, pleiotrophin | |
| This antibody was generated using a synthetic peptide of P21246, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title