missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pleiotrophin/PTN Antibody (CL15628) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43677-25ul
This item is not returnable.
View return policy
Description
Pleiotrophin/PTN Monoclonal antibody specifically detects Pleiotrophin/PTN in Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Pleiotrophin/PTN | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200 | |
| HARP, HBBM, HB-GAM, HBGF8, HBNF, HBNF1, HBNF-1, Heparin-binding brain mitogen, NEGF1HBGF-8, neurite growth-promoting factor 1, neurite growth-promoting factor1), OSF-1, Osteoblast-specific factor 1, pleiotrophin | |
| This antibody was generated using a synthetic peptide of P21246, with the exact immunogen sequence remaining proprietary. | |
| 25 μL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 5764 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL15628 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction